Anti-STOML2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-8861R-A750
BOSSBS-8861R-A750EA 893 EUR
BOSSBS-8861R-A750
Anti-STOML2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
STOML2 is a 356 amino acid member of the mec-2 family of proteins. Expressed ubiquitously at low levels, STOML2 is highly expressed in heart, liver and pancreas. STOML2 is localised to the cytoplasm with some distribution on the membrane. STOML2 was first identified as an overexpressed protein in human endometrial adenocarcinoma. Changes in cell growth in samples with different levels of STOML2 indicate that STOML2 could play a role in endometrial tumorigenesis. STOML2 is also thought to play a role in regulating ion channel conductances or the organisation of sphingolipid and cholesterol-More Product Information