Anti-ITPR1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8869R-CY5.5
BOSSBS-8869R-CY5.5EA 900 EUR
BOSSBS-8869R-CY5.5
Anti-ITPR1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITPR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR