Anti-Vibrio Parahemolyticus Polar flagellin B/D Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-8876R-A750
BOSSBS-8876R-A750EA 893 EUR
BOSSBS-8876R-A750
Anti-Vibrio Parahemolyticus Polar flagellin B/D Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Flagellin is the subunit protein which polymerises to form the filaments of bacterial flagella. FlaB/D is not essential for polar flagellar synthesis and swimming motility. Homopolymer of FlaB/D is not able to form a functional filament.

Type: Primary
Antigen: Vibrio Parahemolyticus Polar flagellin B/D
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/378
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria; Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR