Anti-SARM1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-9001R-CY5
BOSSBS-9001R-CY5EA 900 EUR
BOSSBS-9001R-CY5
Anti-SARM1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Negative regulator of MYD88- and TRIF-dependent toll-like receptor signaling pathway which plays a pivotal role in activating axonal degeneration following injury. Promotes Wallerian degeneration an injury-induced axonal death pathway which involves degeneration of an axon distal to the injury site. Can activate neuronal death in response to stress. Regulates dendritic arborization through the MAPK4-JNK pathway. Involved in innate immune response. Inhibits both TICAM1/TRIF- and MYD88-dependent activation of JUN/AP-1, TRIF-dependent activation of NF-kappa-B and IRF3, and the phosphorylation of MAPK14/p38.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SARM1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR