Anti-EFHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-9015R-A488
BOSSBS-9015R-A488EA 893 EUR
BOSSBS-9015R-A488
Anti-EFHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The EF-hand domain is a twelve amino acid loop motif that is commonly found in proteins that participate in calcium-binding events within the cell. EF-hand domains generally exist in a pair that, together, form a stable four-helix bundle that enables the binding of calcium ions. Swiprosin-2, also known as EFHD1 (EF-hand domain-containing protein D1), SWS2, PP3051 or MST133, is a 239 amino acid protein that contains two EF-hand domains and is expressed in a wide variety of tissues, including brain, liver, heart, kidney, testis, ovaries and spleen. Expression of Swiprosin-2, a possible calcium-binding protein, is upregulated during neuronal differentiation, suggesting a role for Swiprosin-2 in brain development and maturation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EFHD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR