Anti-IQCE Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9018R-A555
BOSSBS-9018R-A555EA 893 EUR
BOSSBS-9018R-A555
Anti-IQCE Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Chromosome 7 is about 158 milllion bases long, encodes over 1000 genes and makes up about 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IQCE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR