Anti-ISLR2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-9025R-CY3
BOSSBS-9025R-CY3EA 900 EUR
BOSSBS-9025R-CY3
Anti-ISLR2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. ISLR2 (immunoglobulin superfamily containing leucine-rich repeat 2), also known as LINX (leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein), is a 745 amino acid single-pass membrane protein that contains five LRR repeats, one Ig-like (immunoglobulin-like) domain, a LRRCT domain and one LRRNT domain. ISLR2 exists as a homomultimer and is essential for axon extension during neural development. The gene encoding ISLR2 maps to human chromosome 15q24.1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ISLR2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR