Anti-HPRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-9026R-A647
BOSSBS-9026R-A647EA 893 EUR
BOSSBS-9026R-A647
Anti-HPRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The protein encoded by this gene is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway. Mutations in this gene result in Lesch-Nyhan syndrome or gout.[provided by RefSeq, Jun 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HPRT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat