Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-9044R-A647
BOSSBS-9044R-A647EA 893 EUR
BOSSBS-9044R-A647
Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Essential ion channel and serine/threonine-protein kinase. Divalent cation channel permeable to calcium and magnesium. Has a central role in magnesium ion homeostasis and in the regulation of anoxic neuronal cell death. Involved in TNF-induced necroptosis downstream of MLKL by mediating calcium influx. The kinase activity is essential for the channel function. May be involved in a fundamental process that adjusts plasma membrane divalent cation fluxes according to the metabolic state of the cell. Phosphorylates annexin A1 (ANXA1).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat