Anti-AKIRIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-9082R-A647
BOSSBS-9082R-A647EA 893 EUR
BOSSBS-9082R-A647
Anti-AKIRIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Required for the innate immune response. Downstream effector of the Toll-like receptor (TLR), TNF and IL-1 beta signaling pathways leading to the production of IL-6. Forms a complex with YWHAB that acts to repress transcription of DUSP1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AKIRIN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR