Anti-AHNAK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-9094R-A555
BOSSBS-9094R-A555EA 893 EUR
BOSSBS-9094R-A555
Anti-AHNAK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
AHNAK2 contains 1 PDZ (DHR) domain. There are 3 named isoforms of AHNAK2 produced by alternative splicing. AHNAK2 is a 600-kDa protein composed of a large number of highly conserved repeat segments. AHNAKs are a class of giant propeller-like proteins that associate with calcium channel proteins of cardiomyocytes and other cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AHNAK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat