Anti-AHNAK2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-9094R-CY7
BOSSBS-9094R-CY7EA 893 EUR
BOSSBS-9094R-CY7
Anti-AHNAK2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
AHNAK2 contains 1 PDZ (DHR) domain. There are 3 named isoforms of AHNAK2 produced by alternative splicing. AHNAK2 is a 600-kDa protein composed of a large number of highly conserved repeat segments. AHNAKs are a class of giant propeller-like proteins that associate with calcium channel proteins of cardiomyocytes and other cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AHNAK2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR