Anti-ANKS1B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-9097R-A680
BOSSBS-9097R-A680EA 893 EUR
BOSSBS-9097R-A680
Anti-ANKS1B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The _-Amyloid protein precursor (AbPP) is a widely expressed transmembrane protein that is processed into the b-Amyloid (Ab) peptide, which accumulates in insoluble plaques in the brain of Alzheimer?s disease patients and AbPP intracellular domain (AID). AID may function as a pro-apoptotic peptide, a regulator of calcium homeostasis and a molecule involved in transcriptional regulation. The AID associated protein 1 (AIDA-1) is highly expressed in the brain and is regulated by AbPP. It interacts with AbPP to play a role in brain development. AIDA-1 also interacts with coilin in Cajal bodies to regulate pre-mRNA splicing.
Type: Primary
Antigen: ANKS1B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 851-1000/1248
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat