Anti-ARTC5 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-9105R-CY5.5
BOSSBS-9105R-CY5.5EA 900 EUR
BOSSBS-9105R-CY5.5
Anti-ARTC5 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Mono-ADP-ribosylation is one of the posttranslational protein modifications regulating cellular metabolism (e.g. nitrogen fixation) in prokaryotes. Mono-ADP-ribosylation is a process in which the ADP-ribose moiety of nicotinamide adenine dinucleotide is transferred to an acceptor amino acid. Five mammalian ADP-ribosyltransferases (ART1-ART5) have been cloned, and each ART is expressed in different tissues. ART5 (ADP-ribosyltransferase 5), also known as Ecto-ADP-ribosyltransferase 5, is a 292 amino acid secretory protein that is expressed in testis, heart, skeletal muscle and lymphoma. Functionally, ART5 is implicatetd to play a role in cell signaling and metabolism cascades. Two isoforms of ART5 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARTC5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR