Anti-CDCA7 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-9128R-CY3
BOSSBS-9128R-CY3EA 1350 EUR
BOSSBS-9128R-CY3
Anti-CDCA7 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CDCA7 is a nuclear protein without a known function, although its high homology with the transcription factor JPO2 or RAM2 suggests that it is also a transcription factor. CDCA7 is one of many target genes regulated by the c-Myc transcription factor. Overexpression of CDCA7 occurs in a significant proportion of cancers and it may play an important role in tumorigenesis. Normally it is periodically expressed in the cell cycle, peaking at the G1 to S phase transition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA7
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
More Product Information
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR