Anti-SERPINB10 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-9211R-CY5
BOSSBS-9211R-CY5EA 900 EUR
BOSSBS-9211R-CY5
Anti-SERPINB10 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
SERPINB10 is a serine proteinase inhibitor of the ovalbumin like B clade of serpins. It was first discovered in human bone marrow in a search for serpins similar to SerpinB6. SERPINB10 was identified in lymphocytes, and was elevated in chronic myeloid leukemia and chronic myelomonocytic leukemia cells. The monocytic cell lines THP-1 and AML-193 also showed elevated SERPINB10 expression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SERPINB10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human