Anti-ARHGAP32 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9296R-A555
BOSSBS-9296R-A555EA 893 EUR
BOSSBS-9296R-A555
Anti-ARHGAP32 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
GTPase-activating protein (GAP) promoting GTP hydrolysis on RHOA, CDC42 and RAC1 small GTPases. May be involved in the differentiation of neuronal cells during the formation of neurite extensions. Involved in NMDA receptor activity-dependent actin reorganization in dendritic spines. May mediate cross-talks between Ras- and Rho-regulated signaling pathways in cell growth regulation. Isoform 2 has higher GAP activity (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARHGAP32
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR