Anti-STIL Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-9303R-A647
BOSSBS-9303R-A647EA 893 EUR
BOSSBS-9303R-A647
Anti-STIL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
TAL1 disruption at 1p32, a common rearrangement in the T-cell acute lymphoblastic leukemia, usually results in the formation of a SCL interrupting locus (SIL)-TAL1 fusion product. SIL is an immediate early gene whose expression is associated with cell proliferation. The Sil protein exhibits ubiquitous expression in hematopoietic cell lines and tissues. However, Sil protein levels remain tightly regulated during the cell cycle, achieving peak levels in mitosis and diminishing on transition to G1 phase. Overexpression of Sil in primary adenocarcinomas predicts metastatic spread, especially in lung tumors with increased mitotic activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STIL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR