Anti-RANBP1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-9322R-CY7
BOSSBS-9322R-CY7EA 900 EUR
BOSSBS-9322R-CY7
Anti-RANBP1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Inhibits GTP exchange on Ran. Forms a Ran-GTP-RANBP1 trimeric complex. Increase GTP hydrolysis induced by the Ran GTPase activating protein RANGAP1. May act in an intracellular signaling pathway which may control the progression through the cell cycle by regulating the transport of protein and nucleic acids across the nuclear membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RANBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR