Anti-MARCH2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-9336R-A350
BOSSBS-9336R-A350EA 893 EUR
BOSSBS-9336R-A350
Anti-MARCH2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). MARCH2 (membrane-associated ring finger (C3HC4) 2), also known as RNF172 or HSPC240, is a 246 amino acid multi-pass membrane protein that localizes to the endoplasmic reticulum and contains one RING-CH-type zinc finger. Expressed in a variety of tissues, MARCH2 functions as an E3 ubiquitin-protein ligase that is thought to mediate the ubiquitination and subsequent degradation of CD71 and B7-2 and may be involved in endosomal protein trafficking.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR