Anti-Proteasome 20S alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-9355R-A750
BOSSBS-9355R-A750EA 893 EUR
BOSSBS-9355R-A750
Anti-Proteasome 20S alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The gene encodes a 246-amino acid polypeptide containing an RNA binding motif, a putative nuclear localisation signal, and phosphorylation sites. The alpha subunits comprises the outer rings of the proteasome. Some alpha subunits contain a functional nuclear localisation signal; proteasomes are found in both the nuclear and cytoplasmic compartments of the cell. Alpha subunits may constitute a physical barrier that limits access of cytosolic proteins into the inner proteolytic chamber.

Type: Primary
Antigen: Proteasome 20S alpha 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/246
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR