Anti-PSMA6 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-9355R-CY5
BOSSBS-9355R-CY5EA 900 EUR
BOSSBS-9355R-CY5
Anti-PSMA6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The gene encodes a 246-amino acid polypeptide containing an RNA binding motif, a putative nuclear localization signal, and phosphorylation sites. The alpha subunits comprises the outer rings of the proteasome. Some alpha subunits contain a functional nuclear localization signal; proteasomes are found in both the nuclear and cytoplasmic compartments of the cell. Alpha subunits may constitute a physical barrier that limits access of cytosolic proteins into the inner proteolytic chamber.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMA6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR