Anti-PSMB6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9358R-A555
BOSSBS-9358R-A555EA 893 EUR
BOSSBS-9358R-A555
Anti-PSMB6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This unit is responsible of the peptidyl glutamyl-like activity. May catalyze basal processing of intracellular antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMB6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR