Anti-Proteasome 20S beta 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-9358R-A750
BOSSBS-9358R-A750EA 893 EUR
BOSSBS-9358R-A750
Anti-Proteasome 20S beta 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The proteasome is a multicatalytic proteinase complex which is characterised by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This unit is responsible of the peptidyl glutamyl-like activity. May catalyze basal processing of intracellular antigens.

Type: Primary
Antigen: Proteasome 20S beta 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/239
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR