Anti-PSMB6 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-9358R-CY5.5
BOSSBS-9358R-CY5.5EA 900 EUR
BOSSBS-9358R-CY5.5
Anti-PSMB6 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This unit is responsible of the peptidyl glutamyl-like activity. May catalyze basal processing of intracellular antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMB6
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR