Anti-MYCBP2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-9375R
BOSSBS-9375REA 725 EUR
BOSSBS-9375R
Anti-MYCBP2 Rabbit Polyclonal Antibody
Antikörper
MYCBP2 belongs to the highwire family. It is a probable E3 ubiquitin-protein ligase which mediates ubiquitination and subsequent proteasomal degradation of target proteins. MYCBP2 may function as a facilitator or regulator of transcriptional activation by MYC and have a role during synaptogenesis. There are two different isoforms.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MYCBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR