Anti-CNOT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9383R-A555
BOSSBS-9383R-A555EA 893 EUR
BOSSBS-9383R-A555
Anti-CNOT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
CNOT2 (CCR4-NOT transcription complex subunit 2) is a ubiquitous protein encoded by the human gene CNOT2. CNOT2 belongs to the CNOT2/3/5 family and is part of the CCR4-NOT complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT2) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. Increased expression of the CNOT2 subunit acts to strongly repress transcription by RNA polymerase II. This repressive effect is mediated by a conserved NOT-Box, which is located at the C-terminus of CNOT2 proteins. Repression by the NOT-Box is sensitive to treatment with the histone deacetylase (HDAC) inhibitor trichostatin A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNOT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR