Anti-CNOT2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-9384R
BOSSBS-9384REA 1090 EUR
BOSSBS-9384R
Anti-CNOT2 Rabbit Polyclonal Antibody
Antikörper
CNOT2 (CCR4-NOT transcription complex subunit 2) is a ubiquitous protein encoded by the human gene CNOT2. CNOT2 belongs to the CNOT2/3/5 family and is part of the CCR4-NOT complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT2) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. Increased expression of the CNOT2 subunit acts to strongly repress transcription by RNA polymerase II. This repressive effect is mediated by a conserved NOT-Box, which is located at the C-terminus of CNOT2 proteins. Repression by the NOT-Box is sensitive to treatment with the histone deacetylase (HDAC) inhibitor trichostatin A.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CNOT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR