Anti-CHAF1A Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-9392R-CY5.5
BOSSBS-9392R-CY5.5EA 900 EUR
BOSSBS-9392R-CY5.5
Anti-CHAF1A Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Core component of the CAF-1 complex, a complex thought to mediate chromatin assembly in DNA replication and DNA repair. Assembles histone octamers onto replicating DNA in vitro. CAF-1 performs the first step of the nucleosome assembly process, bringing newly synthesized histones H3 and H4 to replicating DNA; histones H2A/H2B can bind to this chromatin precursor subsequent to DNA replication to complete the histone octamer. CHAF1A binds to histones H3 and H4. It may play a role in heterochromatin maintenance in proliferating cells by bringing newly synthesized cbx proteins to heterochromaticMore Product Information