Anti-JCHAIN Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-9400R-A488
BOSSBS-9400R-A488EA 893 EUR
BOSSBS-9400R-A488
Anti-JCHAIN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Serves to link two monomer units of either IgM or IgA. In the case of IgM, the J chain-joined dimer is a nucleating unit for the IgM pentamer, and in the case of IgA it induces larger polymers. It also help to bind these immunoglobulins to secretory component.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JCHAIN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR