Anti-5-mC Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-9450R-CY5.5
BOSSBS-9450R-CY5.5EA 900 EUR
BOSSBS-9450R-CY5.5
Anti-5-mC Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Cytidine is a nucleoside formed by a cytosine attached to a ribose ring via a beta-N1-glycosidic bond. DNA is methylated on cytidines by DNA methylases (DNMTs)to generate 5-methylcytidine (5-mC), a potent epigenetics marker and regulator of gene expression. The reverse reaction (cytidine demethylation) starts with its oxidation to hydroxymethyl- (5-hmC), formyl- (5-fC), and carboxy- (5-caC) cytidine. Several enzymes, including the Tet family of proteins have been implicated in cytidine demethylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 5mC
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR