Anti-PDE4DIP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9477R-A555
BOSSBS-9477R-A555EA 893 EUR
BOSSBS-9477R-A555
Anti-PDE4DIP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
May function as an anchor sequestering components of the cAMP-dependent pathway to Golgi and/or centrosomes.Myomegalin, is a 2,346 amino acid protein that contains one NBPF domain and localizes to the nucleus, cytoplasm, centrosome and Golgi apparatus. Expressed at high levels in fetal and adult heart and at lower levels in brain and placenta, myomegalin is thought to function as an anchoring protein that sequesters members of the cAMP-dependent pathway to the Golgi and to centrosomes, thereby mediating cAMP pathway dynamics. Translocations in the gene that encodes myomegalin are associated with myeloproliferative disorders (MBDs), a group of diseases caused by an overproduction of blood cells. Myomegalin exists as twelve isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDE4DIP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR