Anti-CMYA2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9477R-A680
BOSSBS-9477R-A680EA 893 EUR
BOSSBS-9477R-A680
Anti-CMYA2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
May function as an anchor sequestering components of the cAMP-dependent pathway to Golgi and/or centrosomes. Myomegalin, is a 2346 amino acid protein that contains one NBPF domain and localizes to the nucleus, cytoplasm, centrosome and Golgi apparatus. Expressed at high levels in fetal and adult heart and at lower levels in brain and placenta, myomegalin is thought to function as an anchoring protein that sequesters members of the cAMP-dependent pathway to the Golgi and to centrosomes, thereby mediating cAMP pathway dynamics. Translocations in the gene that encodes myomegalin are associated with myeloproliferative disorders (MBDs), a group of diseases caused by an overproduction of blood cells. Myomegalin exists as twelve isoforms due to alternative splicing events.

Type: Primary
Antigen: CMYA2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1501-1600/2365
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR