Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-9509R-A680
BOSSBS-9509R-A680EA 893 EUR
BOSSBS-9509R-A680
Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Leucine-rich repeats (LRRs) are 20-29 amino acid motifs that mediate protein-protein interactions. The primary function of these motifs is to provide a versatile structural framework for the formation of these protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervous system development. Several human diseases are associated with mutation in the genes encoding LRR-containing proteins. The leucine-rich repeat-containing protein 3B (LRRC3B), also designated LRP15, is a 259 amino acid protein that contains 3 LRR repeats. The gene encoding LRRC3B is a tumour suppressor gene that is regulated by DNA methylation. Decreased expression of LRRC3B has been shown in colorectal cancer and gastric cancer, making LRRC3B a candidate marker for those cancers.
Type: Primary
Antigen: LRP15
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 181-259/259
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat