Anti-CTH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-9515R-A680
BOSSBS-9515R-A680EA 890 EUR
BOSSBS-9515R-A680
Anti-CTH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
catalyses the last step in the transsulfuration pathway from methionine to cysteine. Has broad substrate specificity. Converts cystathionine to cysteine, ammonia and 2-oxobutanoate. Converts two cysteine molecules to lanthionine and hydrogen sulfide. Can also accept homocysteine as substrate. Specificity depends on the levels of the endogenous substrates. Generates the endogenous Signalling molecule hydrogen sulfide (H2S), and so contributes to the regulation of blood pressure. Defects in CTH are the cause of cystathioninuria (CSTNU). It is an autosomal recessive phenotype characterised by abnormal accumulation of plasma cystathionine, leading to increased urinary excretion.
Type: Primary
Antigen: CTH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/405
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat