Anti-CTH Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-9515R-CY7
BOSSBS-9515R-CY7EA 897 EUR
BOSSBS-9515R-CY7
Anti-CTH Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Catalyzes the last step in the transsulfuration pathway from methionine to cysteine. Has broad substrate specificity. Converts cystathionine to cysteine, ammonia and 2-oxobutanoate. Converts two cysteine molecules to lanthionine and hydrogen sulfide. Can also accept homocysteine as substrate. Specificity depends on the levels of the endogenous substrates. Generates the endogenous signaling molecule hydrogen sulfide (H2S), and so contributes to the regulation of blood pressure.Defects in CTH are the cause of cystathioninuria (CSTNU). It is an autosomal recessive phenotype characterized by abnormal accumulation of plasma cystathionine, leading to increased urinary excretion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTH
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR