Anti-FECH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-9521R-A647
BOSSBS-9521R-A647EA 893 EUR
BOSSBS-9521R-A647
Anti-FECH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Ferrochelatase catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway, and is localised in the mitochondrion. Defects in ferrochelatase are associated with protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. Porphyrias are a group of inherited or acquired disorders of certain enzymes in the heme biosynthetic pathway (also called porphyrin pathway). They are broadly classified as hepatic porphyrias or erythropoietic porphyrias, based on the site of the overproduction and mainly accumulation of the porphyrins (or their chemical precursors). They manifest with either skin problems, or neurological complications, or occasionally both.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FECH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat