Anti-FECH Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-9521R-CY3
BOSSBS-9521R-CY3EA 900 EUR
BOSSBS-9521R-CY3
Anti-FECH Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Ferrochelatase catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway, and is localised in the mitochondrion. Defects in ferrochelatase are associated with protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. Porphyrias are a group of inherited or acquired disorders of certain enzymes in the heme biosynthetic pathway (also called porphyrin pathway). They are broadly classified as hepatic porphyrias or erythropoietic porphyrias, based on the site of the overproduction and mainly accumulation of the porphyrins (or their chemical precursors). They manifest with either skin problems, or neurological complications, or occasionally both.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FECH
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR