Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-9546R-A750
BOSSBS-9546R-A750EA 1350 EUR
BOSSBS-9546R-A750
Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
FIBCD1 is a 461 amino acid single-pass membrane protein that contains one fibrinogen C-terminal domain and exists as multiple alternatively spliced isoforms. The gene encoding FIBCD1 maps to human chromosome 9, which contains 145 million base pairs, comprises 4% of the human genome and encodes nearly 900 genes.
Type: Primary
Antigen: FIBCD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-461/461
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat