Anti-FIBCD1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-9546R-CY5
BOSSBS-9546R-CY5EA 1350 EUR
BOSSBS-9546R-CY5
Anti-FIBCD1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
FIBCD1 is a 461 amino acid single-pass membrane protein that contains one fibrinogen C-terminal domain and exists as multiple alternatively spliced isoforms. The gene encoding FIBCD1 maps to human chromosome 9, which contains 145 million base pairs, comprises 4% of the human genome and encodes nearly 900 genes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FIBCD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat