Mammalian protein farnesyl transferases are heterodimeric proteins containing two nonidentical Alpha and beta subunits that attach farnesyl residues to a cysteine at the fourth position from the COOH terminus of several proteins, including nuclear lamins and p21Ras proteins. The natural substrates contain the Cys-A-A-Xaa recognition sequence, where the A residues are aliphatic and Xaa represents methionine, serine, glutamine or cysteine. The purified farnesyl transferase is an a-b heterodimer. The beta subunit, which is known as FT beta, CAAX farnesyltransferase subunit beta, or Ras proteins prenyltransferase subunit beta, is a 437 amino acid protein that contains five PFTB repeats and binds the peptide substrate. The Alpha subunit is suspected to participate in formation of a stable complex with the substrate farnesyl pyrophosphate.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PGGT1B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat