Anti-SPAG17 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-9596R-CY7
BOSSBS-9596R-CY7EA 900 EUR
BOSSBS-9596R-CY7
Anti-SPAG17 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
SPAG17 (sperm associated antigen 17), also known as PF6, is a 2,223 amino acid cytoplamic protein that colocalizes with SPAG6 to microtubles. Highly expressed in testis and in organs that contain cilia-bearing cells including brain, oviduct, lung, and uterus, SPAG17 may be important for the structural integrity of the central apparatus of the sperm axoneme. SPAG17 contains two LRR (leucine-rich) repeats and may also participate in flagellar motility and male fertility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPAG17
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR