Anti-DUX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-9664R-A488
BOSSBS-9664R-A488EA 893 EUR
BOSSBS-9664R-A488
Anti-DUX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DUX3 belongs to the paired homeobox family and contains 2 homeobox DNA-binding domains. It expressed in hepatoma Hep3B cells. DUX genes are present in 3.3-kilobase elements, a tandem repeat family scattered in the genome found on the short arms of all acrocentric chromosomes as well as on several other chromosomes. May be functional despite lack of introns and of a poly(A) addition signal. Two different isoforms are known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR