Anti-DUX3 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-9664R-CY5
BOSSBS-9664R-CY5EA 900 EUR
BOSSBS-9664R-CY5
Anti-DUX3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DUX3 belongs to the paired homeobox family and contains 2 homeobox DNA-binding domains. It expressed in hepatoma Hep3B cells. DUX genes are present in 3.3-kilobase elements, a tandem repeat family scattered in the genome found on the short arms of all acrocentric chromosomes as well as on several other chromosomes. May be functional despite lack of introns and of a poly(A) addition signal. Two different isoforms are known.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUX3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human