Anti-DENND2D Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-9668R-A350
BOSSBS-9668R-A350EA 893 EUR
BOSSBS-9668R-A350
Anti-DENND2D Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Guanine nucleotide exchange factor (GEF) which may activate RAB9A and RAB9B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DENND2D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR