Anti-NOL1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-9710R-CY5
BOSSBS-9710R-CY5EA 900 EUR
BOSSBS-9710R-CY5
Anti-NOL1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Proliferation-associated nucleolar antigen is expressed in mid G1 phase with peak level during the S phase and a rapid degradation during late mitosis. Its expression in breast carcinoma is correlated with patient prognosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat