Anti-CNTD1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-9739R-CY3
BOSSBS-9739R-CY3EA 900 EUR
BOSSBS-9739R-CY3
Anti-CNTD1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CNTD

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNTD1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR