Anti-C1orf80 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-9802R-A750
BOSSBS-9802R-A750EA 893 EUR
BOSSBS-9802R-A750
Anti-C1orf80 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
AIDA, also known as C1orf80, is a 306 amino acid protein that belongs to the axin interactor family. Expressed in a variety of tissues, including skeletal muscle and heart, AIDA functions as a ventralizing factor during embryogenesis, disrupting Axin homodimerization and inhibiting Axin-mediated JNK activation. Axin, a scaffold protein, is important for both JNK Signalling and the canonical Wnt pathway, two processes that play an essential role in embryonic dorsoventral patterning. Disruption of Axin by AIDA results in the negative regulation of JNK and Wnt Signalling, thereby affecting embryonic developmental events. Three isoforms of AIDA exist due to alternative splicing events.

Type: Primary
Antigen: C1orf80
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 221-306/306
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR