Anti-C3orf21/XXYLT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-9826R-A680
BOSSBS-9826R-A680EA 893 EUR
BOSSBS-9826R-A680
Anti-C3orf21/XXYLT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Alpha-1,3-xylosyltransferase, which elongates the O-linked xylose-glucose disaccharide attached to EGF-like repeats in the extracellular domain of Notch proteins by catalyzing the addition of the second xylose.
Type: Primary
Antigen: C3orf21/XXYLT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/393
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others