Anti-Cyclophilin 40 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-9878R-A750
BOSSBS-9878R-A750EA 893 EUR
BOSSBS-9878R-A750
Anti-Cyclophilin 40 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Cyclophilins are conserved, ubiquitous and abundant cytosolic peptidyl-prolyl cis-trans isomerases that accelerate the isomerization of XaaPro peptide bonds and the refolding of proteins. Human cyclophilin A (CyPA), an intracellular protein of 165 amino acids, is the target of cyclosporin A (CsA) and is encoded by a single unique gene conserved from yeast to humans. Cyclophilin B (CyPB) is secreted in biological fluids such as blood or milk and binds to a specific receptor present on the human lymphoblastic cell line Jurkat and on human peripheral blood lymphocytes. Cyclophilin D (CyP40) is a widely expressed cytoplasmic protein that catalyses the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. It is a widely expressed cytoplasmic protein.

Type: Primary
Antigen: Cyclophilin 40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 281-370/370
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR